accessibility menu, dialog, popup

UserName

OOHOH

CAS RN 69985-32-6

IUPAC Name: 4-(benzyloxy)butane-1,2-diol
Synonyms: 4-(benzyloxy)butane-1,2-diol
Molecular Weight (g/mol): 196.25
Molecular Formula: C11H16O3
InChi Key: TVRPDIKPMQUOSL-UHFFFAOYNA-N
SMILES: OCC(O)CCOCC1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results