missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-3-Pyrrolidinol 97.0+%, TCI America™
Supplier: TCI America P166725G
Specifications
| (S)-3-Pyrrolidinol | |
| White-Yellow | |
| C4H10NO | |
| 25 g | |
| JHHZLHWJQPUNKB-BYPYZUCNSA-O | |
| (3S)-3-hydroxypyrrolidin-1-ium | |
| 2733874 | |
| ≥97.0% (GC,T) |
| 100243-39-8 | |
| 110°C | |
| MFCD00192426 | |
| s-pyrrolidin-3-ol, s-3-hydroxypyrrolidine, 3s-pyrrolidin-3-ol, s-3-pyrrolidinol, s---3-pyrrolidinol, 3-pyrrolidinol, 3s, 3s-3-pyrrolidinol, 3s-3-hydroxypyrrolidine, s-3-hydroxy-pyrrolidine, s---3-hydroxypyrrolidine | |
| O[C@H]1CC[NH2+]C1 | |
| 88.13 | |
| 87.12 | |
| Crystalline Lumps |
Chemical Identifiers
| 100243-39-8 | |
| 88.13 | |
| JHHZLHWJQPUNKB-BYPYZUCNSA-O | |
| 2733874 | |
| O[C@H]1CC[NH2+]C1 |
| C4H10NO | |
| MFCD00192426 | |
| s-pyrrolidin-3-ol, s-3-hydroxypyrrolidine, 3s-pyrrolidin-3-ol, s-3-pyrrolidinol, s---3-pyrrolidinol, 3-pyrrolidinol, 3s, 3s-3-pyrrolidinol, 3s-3-hydroxypyrrolidine, s-3-hydroxy-pyrrolidine, s---3-hydroxypyrrolidine | |
| (3S)-3-hydroxypyrrolidin-1-ium |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator