accessibility menu, dialog, popup

UserName

OH(S)H2N

CAS RN 100243-39-8

IUPAC Name: (3S)-3-hydroxypyrrolidin-1-ium
Synonyms: (3S)-3-hydroxypyrrolidin-1-ium
Molecular Weight (g/mol): 88.13
Molecular Formula: C4H10NO
InChi Key: JHHZLHWJQPUNKB-BYPYZUCNSA-O
SMILES: O[C@H]1CC[NH2+]C1
Boiling Point 
  • (1)
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)