missing translation for 'onlineSavingsMsg'
Learn More

(S)-3-Pyrrolidinol 97.0+%, TCI America™

Catalog No. p-7135197
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P16671G 1 g
P16675G 5 g
P166725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P16671G Supplier TCI America Supplier No. P16671G
Only null left

Chemical Identifiers

CAS 100243-39-8
Molecular Formula C4H10NO
Molecular Weight (g/mol) 88.13
MDL Number MFCD00192426
InChI Key JHHZLHWJQPUNKB-BYPYZUCNSA-O
Synonym s-pyrrolidin-3-ol, s-3-hydroxypyrrolidine, 3s-pyrrolidin-3-ol, s-3-pyrrolidinol, s---3-pyrrolidinol, 3-pyrrolidinol, 3s, 3s-3-pyrrolidinol, 3s-3-hydroxypyrrolidine, s-3-hydroxy-pyrrolidine, s---3-hydroxypyrrolidine
PubChem CID 2733874
IUPAC Name (3S)-3-hydroxypyrrolidin-1-ium
SMILES O[C@H]1CC[NH2+]C1

Specifications

Color White-Yellow
Boiling Point 110°C
Quantity 1 g
Formula Weight 87.12
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material (S)-3-Pyrrolidinol
TSCA No
Recommended Storage Refrigerator