missing translation for 'onlineSavingsMsg'
Learn More

(S)-3-Boc-thiazolidine-2-carboxylic acid, 97%

Catalog No. p-7035087
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5279303 1 g
AAH5279306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5279303 Supplier Thermo Scientific Chemicals Supplier No. H5279303
Only null left

CAS: 891192-95-3 | C9H15NO4S | 233.282 g/mol

(S)-3-Boc-thiazolidine-2-carboxylic acid is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-3-Boc-thiazolidine-2-carboxylic acid is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 891192-95-3
Molecular Formula C9H15NO4S
Molecular Weight (g/mol) 233.282
MDL Number MFCD02682342
InChI Key HYAXPNDMEODKHI-LURJTMIESA-N
Synonym s-3-boc-2-thiazolidinecarboxylic acid, s-3-tert-butoxycarbonyl thiazolidine-2-carboxylic acid, 2s-3-tert-butoxycarbonyl-1,3-thiazolidine-2-carboxylic acid, 2,3-thiazolidinedicarboxylic acid, 3-1,1-dimethylethyl ester, 2s, s-3-boc-thiazolidine-2-carboxylic acid, n-boc s thiazolidine-2-carboxylic acid, n-boc-s-thiazolidine-2-carboxylic acid, s-thiazolidine-2,3-dicarboxylic acid 3-tert-butyl ester, 2s-3-tert-butoxy carbonyl-1,3-thiazolidine-2-carboxylic acid, 2s-2,3-thiazolidinedicarboxylic acid, 3-1,1-dimethylethyl ester
PubChem CID 14775612
IUPAC Name (2S)-3-[(2-methylpropan-2-yl)oxycarbonyl]-1,3-thiazolidine-2-carboxylic acid
SMILES CC(C)(C)OC(=O)N1CCSC1C(=O)O

Specifications

Melting Point 90°C to 93°C
Quantity 1 g
Optical Rotation −47° (c=5 in Methanol)
Formula Weight 233.28
Percent Purity 97%
Chemical Name or Material (S)-3-Boc-thiazolidine-2-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only