accessibility menu, dialog, popup

UserName

CAS RN 891192-95-3

IUPAC Name: 3-[(tert-butoxy)carbonyl]-1,3-thiazolidine-2-carboxylic acid
Synonyms: 3-(tert-butoxycarbonyl)-1,3-thiazolidine-2-carboxylic acid
Molecular Weight (g/mol): 233.28
Molecular Formula: C9H15NO4S
InChi Key: HYAXPNDMEODKHI-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)N1CCSC1C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results