missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-3-(Boc-amino)-3-phenylpropionic acid, 95%
CAS: 103365-47-5 | C14H19NO4 | 265.309 g/mol
Supplier: Thermo Scientific Chemicals H5207206
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| (S)-3-(Boc-amino)-3-phenylpropionic acid | |
| C14H19NO4 | |
| 5 g | |
| boc-s-3-amino-3-phenylpropionic acid, s-3-tert-butoxycarbonyl amino-3-phenylpropanoic acid, s-3-n-boc-amino-beta-phenylalanine, s-n-boc-3-amino-3-phenylpropanoic acid, boc-d-beta-phe-oh, boc-d-beta-phenylalanine, s-boc-beta3-phenylalanine, boc-s-phenylalanine, 3s-3-tert-butoxycarbonyl amino-3-phenylpropanoic acid, s-3-boc-amino-3-phenylpropionic acid | |
| CC(C)(C)OC(=O)NC(CC(=O)O)C1=CC=CC=C1 | |
| 265.309 | |
| 265.31 |
| 103365-47-5 | |
| MFCD01860892 | |
| 4695945 | |
| JTNQFJPZRTURSI-NSHDSACASA-N | |
| (3S)-3-[(2-methylpropan-2-yl)oxycarbonylamino]-3-phenylpropanoic acid | |
| 2734490 | |
| 95% |
Chemical Identifiers
| 103365-47-5 | |
| 265.309 | |
| JTNQFJPZRTURSI-NSHDSACASA-N | |
| 2734490 | |
| CC(C)(C)OC(=O)NC(CC(=O)O)C1=CC=CC=C1 |
| C14H19NO4 | |
| MFCD01860892 | |
| boc-s-3-amino-3-phenylpropionic acid, s-3-tert-butoxycarbonyl amino-3-phenylpropanoic acid, s-3-n-boc-amino-beta-phenylalanine, s-n-boc-3-amino-3-phenylpropanoic acid, boc-d-beta-phe-oh, boc-d-beta-phenylalanine, s-boc-beta3-phenylalanine, boc-s-phenylalanine, 3s-3-tert-butoxycarbonyl amino-3-phenylpropanoic acid, s-3-boc-amino-3-phenylpropionic acid | |
| (3S)-3-[(2-methylpropan-2-yl)oxycarbonylamino]-3-phenylpropanoic acid |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only