missing translation for 'onlineSavingsMsg'
Learn More

(S)-3-(Boc-amino)-3-phenylpropionic acid, 95%

Catalog No. p-7034389
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Catalog No. Quantity
AAH52072MD 250 mg
AAH5207203 1 g
AAH5207206 5 g
3 options

Catalog No. AAH52072MD

Supplier: Thermo Scientific Chemicals H52072MD

Only null left

CAS: 103365-47-5 | C14H19NO4 | 265.309 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 103365-47-5
Molecular Formula C14H19NO4
Molecular Weight (g/mol) 265.309
MDL Number MFCD01860892
InChI Key JTNQFJPZRTURSI-NSHDSACASA-N
Synonym boc-s-3-amino-3-phenylpropionic acid, s-3-tert-butoxycarbonyl amino-3-phenylpropanoic acid, s-3-n-boc-amino-beta-phenylalanine, s-n-boc-3-amino-3-phenylpropanoic acid, boc-d-beta-phe-oh, boc-d-beta-phenylalanine, s-boc-beta3-phenylalanine, boc-s-phenylalanine, 3s-3-tert-butoxycarbonyl amino-3-phenylpropanoic acid, s-3-boc-amino-3-phenylpropionic acid
PubChem CID 2734490
IUPAC Name (3S)-3-[(2-methylpropan-2-yl)oxycarbonylamino]-3-phenylpropanoic acid
SMILES CC(C)(C)OC(=O)NC(CC(=O)O)C1=CC=CC=C1

Specifications

Quantity 250 mg
Beilstein 4695945
Formula Weight 265.31
Percent Purity 95%
Chemical Name or Material (S)-3-(Boc-amino)-3-phenylpropionic acid
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only