missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ S 2101
Description
S 2101 is a LSD1 inhibitor (IC50 = 990 nM; Ki = 610 nM). Exhibits selectivity for LSD1 over MAO-B and MAO-A (Ki values are 17 and 110 μM, respectively).
Specifications
Specifications
| Chemical Name or Material | (1R,2S)-rel-2-[3,5-Difluoro-2-(phenylmethoxy)phenyl]cycloprpanamine hydrochloride |
| CAS | 1239262-36-2 |
| Quantity | 50 mg |
| Product Line | Tocris |
| Target | Histone Demethylase Inhibitors |
| Molecular Formula | C16H15F2NO.HCl |
| Formula Weight | 311.75 |
| Purity | >98% |