missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-(+)-2-Octanol 98.0+%, TCI America™
$205.05 - $647.43
Chemical Identifiers
| CAS | 6169-06-8 |
|---|---|
| Molecular Formula | C8H18O |
| Molecular Weight (g/mol) | 130.23 |
| MDL Number | MFCD00064283 |
| InChI Key | SJWFXCIHNDVPSH-UHFFFAOYNA-N |
| Synonym | s-+-2-octanol, s-octan-2-ol, s-2-octanol, 2s-octan-2-ol, d +-2-octanol, d-octan-2-ol, 2s-2-octanol, 2-octanol, 2s, l-2-octanol, 2-octanol s |
| PubChem CID | 2723888 |
| ChEBI | CHEBI:37870 |
| IUPAC Name | (2S)-octan-2-ol |
| SMILES | CCCCCCC(C)O |
Chemical Identifiers
| C8H18O | |
| MFCD00064283 | |
| s-+-2-octanol, s-octan-2-ol, s-2-octanol, 2s-octan-2-ol, d +-2-octanol, d-octan-2-ol, 2s-2-octanol, 2-octanol, 2s, l-2-octanol, 2-octanol s | |
| CHEBI:37870 | |
| CCCCCCC(C)O |
Specifications
| 6169-06-8 | |
| C8H18O | |
| s-+-2-octanol, s-octan-2-ol, s-2-octanol, 2s-octan-2-ol, d +-2-octanol, d-octan-2-ol, 2s-2-octanol, 2-octanol, 2s, l-2-octanol, 2-octanol s | |
| CCCCCCC(C)O | |
| 130.23 | |
| CHEBI:37870 | |
| ≥98.0% (GC) | |
| (S)-(+)-2-Octanol |
| Colorless | |
| MFCD00064283 | |
| SJWFXCIHNDVPSH-UHFFFAOYNA-N | |
| (2S)-octan-2-ol | |
| 2723888 | |
| 130.23 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0217
TSCA : Yes