missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-(+)-2-Octanol 98.0+%, TCI America™
Supplier: TCI America O014425ML
| Quantity | 25 mL |
|---|
Chemical Identifiers
| C8H18O | |
| MFCD00064283 | |
| s-+-2-octanol, s-octan-2-ol, s-2-octanol, 2s-octan-2-ol, d +-2-octanol, d-octan-2-ol, 2s-2-octanol, 2-octanol, 2s, l-2-octanol, 2-octanol s | |
| CHEBI:37870 | |
| CCCCCCC(C)O |
Specifications
| (S)-(+)-2-Octanol | |
| Colorless | |
| 25 mL | |
| s-+-2-octanol, s-octan-2-ol, s-2-octanol, 2s-octan-2-ol, d +-2-octanol, d-octan-2-ol, 2s-2-octanol, 2-octanol, 2s, l-2-octanol, 2-octanol s | |
| CCCCCCC(C)O | |
| 130.23 | |
| CHEBI:37870 | |
| ≥98.0% (GC) |
| 6169-06-8 | |
| C8H18O | |
| MFCD00064283 | |
| SJWFXCIHNDVPSH-UHFFFAOYNA-N | |
| (2S)-octan-2-ol | |
| 2723888 | |
| 130.23 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0217
TSCA : Yes