missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2-[Hydroxy(diphenyl)methyl]-1-methylpyrrolidine 98.0+%, TCI America™

Catalog No. p-7130714
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0768100MG 100 mg
H07681G 1 g
H07685G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. H0768100MG Supplier TCI America Supplier No. H0768100MG
Only null left

Chemical Identifiers

CAS 110529-22-1
Molecular Formula C18H22NO+
Molecular Weight (g/mol) 268.38
MDL Number MFCD00145245
InChI Key XIJAGFLYYNXCAB-KRWDZBQOSA-O
Synonym 1-methyl-2alpha-alpha-hydroxybenzhydryl pyrrolidinium, 2s-2-hydroxydiphenylmethyl-1-methylpyrrolidin-1-ium, 2s-1-methylpyrrolidin-1-ium-2-yl-diphenylmethanol
PubChem CID 6950266
IUPAC Name [(2S)-1-methylpyrrolidin-1-ium-2-yl]-diphenylmethanol
SMILES C[NH+]1CCCC1C(C2=CC=CC=C2)(C3=CC=CC=C3)O

Specifications

Melting Point 71°C
Color White-Yellow
Quantity 100 mg
Formula Weight 267.37
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (S)-(+)-2-[Hydroxy(diphenyl)methyl]-1-methylpyrrolidine