Learn More
S-(2-Aminoethyl)isothiourea dihydrobromide, 98%
CAS: 56-10-0 | C3H11Br2N3S | 281.01 g/mol
Supplier: Thermo Scientific Chemicals L1423206
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| S-(2-Aminoethyl)isothiourea dihydrobromide | |
| 190°C to 192°C | |
| C3H11Br2N3S | |
| 5 g | |
| 14,178 | |
| XDVMCVGTDUKDHL-UHFFFAOYSA-N | |
| 2-aminoethyl carbamimidothioate;dihydrobromide | |
| 5940 | |
| 98% |
| 56-10-0 | |
| Odorless | |
| MFCD00037011 | |
| 3911163 | |
| 2-2-aminoethyl isothiourea dihydrobromide, antiradon, antirad, surrectan, ixecur, 2-aminoethyl carbamimidothioate dihydrobromide, usaf xr-31, 2-2-aminoethyl-2-thiopseudourea dihydrobromide, 2-aminoethylisothiouronium dibromide, carbamimidothioic acid, 2-aminoethyl ester, dihydrobromide | |
| C(CSC(=N)N)N.Br.Br | |
| 281.01 | |
| 281.02 |
Chemical Identifiers
| 56-10-0 | |
| 281.01 | |
| XDVMCVGTDUKDHL-UHFFFAOYSA-N | |
| 5940 | |
| C(CSC(=N)N)N.Br.Br |
| C3H11Br2N3S | |
| MFCD00037011 | |
| 2-2-aminoethyl isothiourea dihydrobromide, antiradon, antirad, surrectan, ixecur, 2-aminoethyl carbamimidothioate dihydrobromide, usaf xr-31, 2-2-aminoethyl-2-thiopseudourea dihydrobromide, 2-aminoethylisothiouronium dibromide, carbamimidothioic acid, 2-aminoethyl ester, dihydrobromide | |
| 2-aminoethyl carbamimidothioate;dihydrobromide |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
GHS P Statement
P280f
Wear protective clothing.
Warning
EINECSNumber : 200-257-0
RTECSNumber : UM0175000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only