missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-2-Amino-3-methyl-1,1-diphenyl-1-butanol 98.0+%, TCI America™

Catalog No. A25041G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A25041G 1 g
A25045G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A25041G Supplier TCI America Supplier No. A25041G
Only null left

Chemical Identifiers

CAS 78603-95-9
Molecular Formula C17H21NO
Molecular Weight (g/mol) 255.36
MDL Number MFCD01318248
InChI Key LNQVZZGGOZBOQS-UHFFFAOYNA-N
PubChem CID 854145
IUPAC Name 2-amino-3-methyl-1,1-diphenylbutan-1-ol
SMILES CC(C)C(N)C(O)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 98°C
Color White-Yellow
Quantity 1 g
Formula Weight 255.36
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (S)-(-)-2-Amino-3-methyl-1,1-diphenyl-1-butanol