missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-2-Amino-3,3-dimethyl-1,1-diphenyl-1-butanol 98.0+%, TCI America™

Catalog No. A25031G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A25031G 1 g
A25035G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A25031G Supplier TCI America Supplier No. A25031G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 144054-70-6
Molecular Formula C18H23NO
Molecular Weight (g/mol) 269.388
MDL Number MFCD03427198
InChI Key HZIHDWNOPKIOCK-INIZCTEOSA-N
PubChem CID 10333308
IUPAC Name (2S)-2-amino-3,3-dimethyl-1,1-diphenylbutan-1-ol
SMILES CC(C)(C)C(C(C1=CC=CC=C1)(C2=CC=CC=C2)O)N

Specifications

Melting Point 134°C
Color White
Quantity 1 g
Formula Weight 269.39
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material (S)-(-)-2-Amino-3,3-dimethyl-1,1-diphenyl-1-butanol