accessibility menu, dialog, popup

UserName

CH3CH3H3COHH2N

CAS RN 144054-70-6

IUPAC Name: 2-amino-3,3-dimethyl-1,1-diphenylbutan-1-ol
Synonyms: 2-amino-3,3-dimethyl-1,1-diphenylbutan-1-ol
Molecular Weight (g/mol): 269.39
Molecular Formula: C18H23NO
InChi Key: HZIHDWNOPKIOCK-UHFFFAOYNA-N
SMILES: CC(C)(C)C(N)C(O)(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results