missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2,2,2-Trifluoro-1-(9-anthryl)ethanol 99.0+%, TCI America™

Catalog No. T1521100MG
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T1521100MG 100 mg
T15211G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T1521100MG Supplier TCI America Supplier No. T1521100MG

Chemical Identifiers

CAS 60646-30-2
Molecular Formula C16H11F3O
Molecular Weight (g/mol) 276.26
MDL Number MFCD00062968
InChI Key ICZHJFWIOPYQCA-UHFFFAOYNA-N
Synonym s-+-2,2,2-trifluoro-1-9-anthryl ethanol, s-+-1-9-anthryl-2,2,2-trifluoroethanol, s-1-anthracen-9-yl-2,2,2-trifluoroethanol, s-2,2,2-trifluoro-1-9-anthryl ethanol, 1s-1-anthracen-9-yl-2,2,2-trifluoroethanol, 1-9-anthryl-2,2,2-trifluoroethanol #, iczhjfwiopyqca-hnnxbmfysa, 1s-1-9-anthracenyl-2,2,2-trifluoroethanol, s-+-, a-trifluoromethyl anthracene-9-methanol
PubChem CID 2724111
IUPAC Name 1-(anthracen-9-yl)-2,2,2-trifluoroethan-1-ol
SMILES OC(C1=C2C=CC=CC2=CC2=CC=CC=C12)C(F)(F)F

Specifications

Melting Point 135°C
Quantity 100 mg
Formula Weight 276.26
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (S)-(+)-2,2,2-Trifluoro-1-(9-anthryl)ethanol [e.e. Determination Reagent by NMR]