accessibility menu, dialog, popup

UserName

OHFFF

CAS RN 60646-30-2

IUPAC Name: 1-(anthracen-9-yl)-2,2,2-trifluoroethan-1-ol
Synonyms: pirkle's alcohol
Molecular Weight (g/mol): 276.26
Molecular Formula: C16H11F3O
InChi Key: ICZHJFWIOPYQCA-UHFFFAOYNA-N
SMILES: OC(C1=C2C=CC=CC2=CC2=CC=CC=C12)C(F)(F)F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results