missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-1-Benzyloxy-2-propanol 95.0+%, TCI America™

Catalog No. b41171g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Catalog No. Quantity
B41171G 1 g
B41175G 5 g
2 options

Catalog No. B41171G

Supplier: TCI America B41171G

Only null left

Chemical Identifiers

CAS 85483-97-2
Molecular Formula C10H14O2
Molecular Weight (g/mol) 166.22
MDL Number MFCD07367008
InChI Key KJBPYIUAQLPHJG-VIFPVBQESA-N
Synonym s-+-1-benzyloxy-2-propanol, s-1-benzyloxy propan-2-ol, 2s-1-benzyloxypropan-2-ol, 2s-1-benzyloxy propan-2-ol, 2-propanol, 1-phenylmethoxy-, 2s, 2-propanol,1-phenylmethoxy-, 2s, s-1-o-benzylpropylene glycol, s-1-benzyloxy-2-hydroxypropane, 2s-1-benzyloxy-2-propanol, +-s-1-benzyloxy-2-propanol
PubChem CID 13197475
IUPAC Name (2S)-1-phenylmethoxypropan-2-ol
SMILES CC(COCC1=CC=CC=C1)O

Specifications

Color Colorless
Boiling Point 81°C
Quantity 1 g
Formula Weight 166.22
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material (S)-(+)-1-Benzyloxy-2-propanol