missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-1-Benzyloxy-2-propanol, 96%

Catalog No. p-7040685
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6052303 1 g
AAH6052306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6052303 Supplier Thermo Scientific Chemicals Supplier No. H6052303

CAS: 85483-97-2 | C10H14O2 | 166.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 85483-97-2
Molecular Formula C10H14O2
Molecular Weight (g/mol) 166.22
MDL Number MFCD07367008
InChI Key KJBPYIUAQLPHJG-VIFPVBQESA-N
Synonym s-+-1-benzyloxy-2-propanol, s-1-benzyloxy propan-2-ol, 2s-1-benzyloxypropan-2-ol, 2s-1-benzyloxy propan-2-ol, 2-propanol, 1-phenylmethoxy-, 2s, 2-propanol,1-phenylmethoxy-, 2s, s-1-o-benzylpropylene glycol, s-1-benzyloxy-2-hydroxypropane, 2s-1-benzyloxy-2-propanol, +-s-1-benzyloxy-2-propanol
PubChem CID 13197475
IUPAC Name (2S)-1-phenylmethoxypropan-2-ol
SMILES CC(COCC1=CC=CC=C1)O

Specifications

Density 1.044
Flash Point 108°C (226°F)
Refractive Index 1.51
Quantity 1 g
Optical Rotation +14° (c=1 in Chloroform)
Formula Weight 166.22
Percent Purity 96%
Chemical Name or Material (S)-(+)-1-Benzyloxy-2-propanol
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only