missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-1-(4-Fluorophenyl)ethylamine, ChiPros 99%, ee 99%

CAS: 66399-30-2 | C8H10FN | 139.173 g/mol

Supplier:  Thermo Scientific Chemicals L1912114

 View more versions of this product

Catalog No. AAL1912114


Only null left
Explore available promotions

Description

Description

(S)-(-)-1-(4-Fluorophenyl)ethylamine is used in organic synthesis. It acts as a ligand and form a coordination complex, diacetato-2-O-bis[(S)-1-(4-fluorophenyl)ethylamine-N]palladium(II) by reacting with palladium(II) acetate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-(-)-1-(4-Fluorophenyl)ethylamine is used in organic synthesis. It acts as a ligand and form a coordination complex, diacetato-2-O-bis[(S)-1-(4-fluorophenyl)ethylamine-N]palladium(II) by reacting with palladium(II) acetate.

Solubility
Miscible with dimethyl sulfoxide.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

(S)-(-)-1-(4-Fluorophenyl)ethylamine
-30°C
76°C (22 mmHg)
ee 99%
1.502
25 g
6791614
s-1-4-fluorophenyl ethylamine, a-methylbenzylamine, 1s-1-4-fluorophenyl ethanamine, 1s-1-4-fluorophenyl ethanamine, 1s-1-4-fluorophenyl ethylamine, s---1-4-fluorophenyl ethylamine, s---1-4-fluorophenyl ethylamine, pubchem15218, pubchem15218, s-4-fluoro-, s-4-fluoro-
QGCLEUGNYRXBMZ-LURJTMIESA-N
CC(C1=CC=C(C=C1)F)N
139.173
139.17
66399-30-2
1.03
68°C (154°F)
C8H10FN
MFCD03092999
UN2735
Air Sensitive
Miscible with dimethyl sulfoxide.
−33° (Neat)
(1S)-1-(4-fluorophenyl)ethanamine
2779063
99%

Chemical Identifiers

66399-30-2
139.173
QGCLEUGNYRXBMZ-LURJTMIESA-N
2779063
CC(C1=CC=C(C=C1)F)N
C8H10FN
MFCD03092999
s-1-4-fluorophenyl ethylamine, a-methylbenzylamine, 1s-1-4-fluorophenyl ethanamine, 1s-1-4-fluorophenyl ethanamine, 1s-1-4-fluorophenyl ethylamine, s---1-4-fluorophenyl ethylamine, s---1-4-fluorophenyl ethylamine, pubchem15218, pubchem15218, s-4-fluoro-, s-4-fluoro-
(1S)-1-(4-fluorophenyl)ethanamine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H227
Causes severe skin burns and eye damage.
Combustible liquid.

P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H227-H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

EINECSNumber : 679-261-3

TSCA : No

Recommended Storage : Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only