accessibility menu, dialog, popup

UserName

CAS RN 66399-30-2

IUPAC Name: 1-(4-fluorophenyl)ethan-1-amine
Synonyms: 1-(4-fluorophenyl)ethanamine
Molecular Weight (g/mol): 139.17
Molecular Formula: C8H10FN
InChi Key: QGCLEUGNYRXBMZ-UHFFFAOYNA-N
SMILES: CC(N)C1=CC=C(F)C=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results