missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-1-(4-Fluorophenyl)ethylamine, ChiPros 99%, ee 99%

Catalog No. aal1912106
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
AAL1912106 5 g
AAL1912114 25 g
2 options

Catalog No. AAL1912106

Supplier: Thermo Scientific Chemicals L1912106

Only null left

CAS: 66399-30-2 | C8H10FN | 139.173 g/mol

(S)-(-)-1-(4-Fluorophenyl)ethylamine is used in organic synthesis. It acts as a ligand and form a coordination complex, diacetato-2-O-bis[(S)-1-(4-fluorophenyl)ethylamine-N]palladium(II) by reacting with palladium(II) acetate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-(-)-1-(4-Fluorophenyl)ethylamine is used in organic synthesis. It acts as a ligand and form a coordination complex, diacetato-2-O-bis[(S)-1-(4-fluorophenyl)ethylamine-N]palladium(II) by reacting with palladium(II) acetate.

Solubility
Miscible with dimethyl sulfoxide.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 66399-30-2
Molecular Formula C8H10FN
Molecular Weight (g/mol) 139.173
MDL Number MFCD03092999
InChI Key QGCLEUGNYRXBMZ-LURJTMIESA-N
Synonym s-1-4-fluorophenyl ethylamine, s-1-4-fluorophenyl ethanamine, 1s-1-4-fluorophenyl ethanamine, 1s-1-4-fluorophenyl ethylamine, s-1-4-fluoro-phenyl-ethylamine, s---1-4-fluorophenyl ethylamine, 1s-1-4-fluorophenyl ethan-1-amine, pubchem15218, 5-1-4-fluorophenyl ethanamine, s-4-fluoro-, a-methylbenzylamine
PubChem CID 2779063
IUPAC Name (1S)-1-(4-fluorophenyl)ethanamine
SMILES CC(C1=CC=C(C=C1)F)N

Specifications

Melting Point -30°C
Density 1.03
Boiling Point 76°C (22 mmHg)
Flash Point 68°C (154°F)
Refractive Index 1.502
Quantity 5 g
UN Number UN2735
Beilstein 6791614
Sensitivity Air Sensitive
Solubility Information Miscible with dimethyl sulfoxide.
Optical Rotation −33° (Neat)
Formula Weight 139.17
Percent Purity 99%
Assay ee 99%
Chemical Name or Material (S)-(-)-1-(4-Fluorophenyl)ethylamine
Show More Show Less
Hazard Category H227-H314-H335
Hazard Statement GHS H Statement
H314-H227
Causes severe skin burns and eye damage.
Combustible liquid.
Precautionary Statement P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber 679-261-3
TSCA No
Recommended Storage Store under Nitrogen

RUO – Research Use Only