missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic Acid 98.0+%, TCI America™

Supplier:  TCI America T151525G

 View more versions of this product

Catalog No. T151525G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

(S)-(-)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic Acid
White-Yellow
MFCD00144533
l-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, s-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, h-tic-oh, l-porretine, 3s-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, s-1,2,3,4-tetrahydro-3-isoquinolinecarboxylic acid, unii-737g46u1np, s---1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, 3s-1,2,3,4-tetrahydro-isoquinoline-3-carboxylic acid
OC(=O)[C@@H]1CC2=CC=CC=C2CN1
177.20
177.20
Crystalline Powder
74163-81-8
C10H11NO2
25 g
BWKMGYQJPOAASG-VIFPVBQESA-N
(3S)-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid
2733226
≥98.0% (T)

Chemical Identifiers

74163-81-8
177.20
BWKMGYQJPOAASG-VIFPVBQESA-N
2733226
OC(=O)[C@@H]1CC2=CC=CC=C2CN1
C10H11NO2
MFCD00144533
l-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, s-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, h-tic-oh, l-porretine, 3s-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, s-1,2,3,4-tetrahydro-3-isoquinolinecarboxylic acid, unii-737g46u1np, s---1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, 3s-1,2,3,4-tetrahydro-isoquinoline-3-carboxylic acid
(3S)-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions