missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic Acid 98.0+%, TCI America™

Catalog No. p-7136814
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T15155G 5 g
T151525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T15155G Supplier TCI America Supplier No. T15155G
Only null left

Chemical Identifiers

CAS 74163-81-8
Molecular Formula C10H11NO2
Molecular Weight (g/mol) 177.20
MDL Number MFCD00144533
InChI Key BWKMGYQJPOAASG-VIFPVBQESA-N
Synonym l-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, s-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, h-tic-oh, l-porretine, 3s-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, s-1,2,3,4-tetrahydro-3-isoquinolinecarboxylic acid, unii-737g46u1np, s---1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, 3s-1,2,3,4-tetrahydro-isoquinoline-3-carboxylic acid
PubChem CID 2733226
IUPAC Name (3S)-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid
SMILES OC(=O)[C@@H]1CC2=CC=CC=C2CN1

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 177.20
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (S)-(-)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic Acid