accessibility menu, dialog, popup

UserName

CAS RN 74163-81-8

IUPAC Name: (3S)-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid
Synonyms: (3S)-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid
Molecular Weight (g/mol): 177.2
Molecular Formula: C10H11NO2
InChi Key: BWKMGYQJPOAASG-VIFPVBQESA-N
SMILES: OC(=O)[C@@H]1CC2=CC=CC=C2CN1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)