Learn More
(S)-(-)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid, 97%
CAS: 74163-81-8 | C10H11NO2 | 177.20 g/mol
Supplier: Thermo Scientific Chemicals L1442203
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| (S)-(-)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid | |
| >300°C (decomposition) | |
| MFCD00144533 | |
| 4842199 | |
| BWKMGYQJPOAASG-VIFPVBQESA-N | |
| OC(=O)[C@@H]1CC2=CC=CC=C2CN1 | |
| 177.20 | |
| 177.2 |
| 74163-81-8 | |
| C10H11NO2 | |
| 1 g | |
| l-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, s-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, h-tic-oh, l-porretine, 3s-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, s-1,2,3,4-tetrahydro-3-isoquinolinecarboxylic acid, unii-737g46u1np, s---1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, 3s-1,2,3,4-tetrahydro-isoquinoline-3-carboxylic acid | |
| −163° (c=2 in 1N NaOH) | |
| (3S)-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid | |
| 2733226 | |
| 97% |
Chemical Identifiers
| 74163-81-8 | |
| 177.20 | |
| BWKMGYQJPOAASG-VIFPVBQESA-N | |
| 2733226 | |
| OC(=O)[C@@H]1CC2=CC=CC=C2CN1 |
| C10H11NO2 | |
| MFCD00144533 | |
| l-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, s-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, h-tic-oh, l-porretine, 3s-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, s-1,2,3,4-tetrahydro-3-isoquinolinecarboxylic acid, unii-737g46u1np, s---1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, 3s-1,2,3,4-tetrahydro-isoquinoline-3-carboxylic acid | |
| (3S)-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only