missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid, 97%

Catalog No. p-7046463
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1442203 1 g
AAL1442206 5 g
AAL1442214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1442203 Supplier Thermo Scientific Chemicals Supplier No. L1442203

CAS: 74163-81-8 | C10H11NO2 | 177.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 74163-81-8
Molecular Formula C10H11NO2
Molecular Weight (g/mol) 177.20
MDL Number MFCD00144533
InChI Key BWKMGYQJPOAASG-VIFPVBQESA-N
Synonym l-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, s-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, h-tic-oh, l-porretine, 3s-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, s-1,2,3,4-tetrahydro-3-isoquinolinecarboxylic acid, unii-737g46u1np, s---1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, 3s-1,2,3,4-tetrahydro-isoquinoline-3-carboxylic acid
PubChem CID 2733226
IUPAC Name (3S)-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid
SMILES OC(=O)[C@@H]1CC2=CC=CC=C2CN1

Specifications

Melting Point >300°C (decomposition)
Quantity 1 g
Beilstein 4842199
Optical Rotation −163° (c=2 in 1N NaOH)
Formula Weight 177.2
Percent Purity 97%
Chemical Name or Material (S)-(-)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only