missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-(+)-1,2,3,4-Tetrahydro-1-naphthylamine 98.0+%, TCI America™
Supplier: TCI America T287825G
Specifications
| (S)-(+)-1,2,3,4-Tetrahydro-1-naphthylamine | |
| 77°C | |
| MFCD00671630 | |
| s-1,2,3,4-tetrahydronaphthalen-1-amine, s-1,2,3,4-tetrahydro-1-naphthylamine, s-+-1,2,3,4-tetrahydro-1-naphthylamine, 1s-1,2,3,4-tetrahydronaphthalen-1-amine, s-1-amino-1,2,3,4-tetrahydronaphthalene, s-+-1-aminotetraline, chembl39537, 1-naphthalenamine, 1,2,3,4-tetrahydro-, 1s, s-1,2,3,4-tetrahydro-naphthalen-1-ylamine, s-1-amino-1,2,3,4-tetrahydro-naphthalene | |
| C1CC(C2=CC=CC=C2C1)N | |
| 147.221 | |
| 147.22 | |
| Liquid |
| 23357-52-0 | |
| C10H13N | |
| 25 g | |
| JRZGPXSSNPTNMA-JTQLQIEISA-N | |
| (1S)-1,2,3,4-tetrahydronaphthalen-1-amine | |
| 7058074 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 23357-52-0 | |
| 147.221 | |
| JRZGPXSSNPTNMA-JTQLQIEISA-N | |
| 7058074 | |
| C1CC(C2=CC=CC=C2C1)N |
| C10H13N | |
| MFCD00671630 | |
| s-1,2,3,4-tetrahydronaphthalen-1-amine, s-1,2,3,4-tetrahydro-1-naphthylamine, s-+-1,2,3,4-tetrahydro-1-naphthylamine, 1s-1,2,3,4-tetrahydronaphthalen-1-amine, s-1-amino-1,2,3,4-tetrahydronaphthalene, s-+-1-aminotetraline, chembl39537, 1-naphthalenamine, 1,2,3,4-tetrahydro-, 1s, s-1,2,3,4-tetrahydro-naphthalen-1-ylamine, s-1-amino-1,2,3,4-tetrahydro-naphthalene | |
| (1S)-1,2,3,4-tetrahydronaphthalen-1-amine |
Safety and Handling
TSCA : No