missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-1,2,3,4-Tetrahydro-1-naphthylamine 98.0+%, TCI America™

Catalog No. p-7137463
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T28785G 5 g
T287825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T28785G Supplier TCI America Supplier No. T28785G

Chemical Identifiers

CAS 23357-52-0
Molecular Formula C10H13N
Molecular Weight (g/mol) 147.221
MDL Number MFCD00671630
InChI Key JRZGPXSSNPTNMA-JTQLQIEISA-N
Synonym s-1,2,3,4-tetrahydronaphthalen-1-amine, s-1,2,3,4-tetrahydro-1-naphthylamine, s-+-1,2,3,4-tetrahydro-1-naphthylamine, 1s-1,2,3,4-tetrahydronaphthalen-1-amine, s-1-amino-1,2,3,4-tetrahydronaphthalene, s-+-1-aminotetraline, chembl39537, 1-naphthalenamine, 1,2,3,4-tetrahydro-, 1s, s-1,2,3,4-tetrahydro-naphthalen-1-ylamine, s-1-amino-1,2,3,4-tetrahydro-naphthalene
PubChem CID 7058074
IUPAC Name (1S)-1,2,3,4-tetrahydronaphthalen-1-amine
SMILES C1CC(C2=CC=CC=C2C1)N

Specifications

Boiling Point 77°C
Quantity 5 g
Formula Weight 147.22
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material (S)-(+)-1,2,3,4-Tetrahydro-1-naphthylamine