missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-(+)-1,2,3,4-Tetrahydro-1-naphthol 98.0+%, TCI America™
$218.10 - $1444.65
Chemical Identifiers
| CAS | 53732-47-1 |
|---|---|
| Molecular Formula | C10H12O |
| Molecular Weight (g/mol) | 148.205 |
| MDL Number | MFCD00063006 |
| InChI Key | JAAJQSRLGAYGKZ-JTQLQIEISA-N |
| Synonym | (S)-(+)-alpha-Tetralol |
| PubChem CID | 7058029 |
| IUPAC Name | (1S)-1,2,3,4-tetrahydronaphthalen-1-ol |
| SMILES | C1CC(C2=CC=CC=C2C1)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
T2360100MG
|
TCI America
T2360100MG |
100 mg |
Each for $218.10
|
|
|||||
|
T23601G
|
TCI America
T23601G |
1 g |
Each for $1,444.65
|
|
|||||
Chemical Identifiers
| 53732-47-1 | |
| 148.205 | |
| JAAJQSRLGAYGKZ-JTQLQIEISA-N | |
| 7058029 | |
| C1CC(C2=CC=CC=C2C1)O |
| C10H12O | |
| MFCD00063006 | |
| (S)-(+)-alpha-Tetralol | |
| (1S)-1,2,3,4-tetrahydronaphthalen-1-ol |
Specifications
| 53732-47-1 | |
| White-Yellow | |
| C10H12O | |
| 100 mg | |
| JAAJQSRLGAYGKZ-JTQLQIEISA-N | |
| (1S)-1,2,3,4-tetrahydronaphthalen-1-ol | |
| 7058029 | |
| ≥98.0% (GC) | |
| (S)-(+)-1,2,3,4-Tetrahydro-1-naphthol |
| 41°C | |
| 92°C | |
| MFCD00063006 | |
| (S)-(+)-alpha-Tetralol | |
| C1CC(C2=CC=CC=C2C1)O | |
| 148.205 | |
| 148.21 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : QL5075000
TSCA : No