CAS RN 53732-47-1
CAS RN 53732-47-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(S)-(+)-1,2,3,4-Tetrahydro-1-naphthol 98.0+%, TCI America™
CAS: 53732-47-1 Molecular Formula: C10H12O Molecular Weight (g/mol): 148.205 MDL Number: MFCD00063006 InChI Key: JAAJQSRLGAYGKZ-JTQLQIEISA-N Synonym: (S)-(+)-alpha-Tetralol PubChem CID: 7058029 IUPAC Name: (1S)-1,2,3,4-tetrahydronaphthalen-1-ol SMILES: C1CC(C2=CC=CC=C2C1)O