missing translation for 'onlineSavingsMsg'
Learn More

(S)-1,2,3,4-Tetrahydro-1-naphthoic acid, 98%

Catalog No. p-7030076
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH27612MD 250 mg
AAH2761203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH27612MD Supplier Thermo Scientific Chemicals Supplier No. H27612MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 85977-52-2 | C11H12O2 | 176.215 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 85977-52-2
Molecular Formula C11H12O2
Molecular Weight (g/mol) 176.215
MDL Number MFCD07368365
InChI Key VDLWTJCSPSUGOA-JTQLQIEISA-N
Synonym s-1,2,3,4-tetrahydro-1-naphthoic acid, s-1,2,3,4-tetrahedro-naphthoic acid, 1s-1,2,3,4-tetrahydronaphthalene-1-carboxylic acid, s-1,2,3,4-tetrahydro-naphthoic acid, 1s-tetralin-1-carboxylic acid, s---1,2,3,4-tetrahydronaphthoic acid, s-1,2,3,4-tetrahydronaphthalene-1-carboxylic acid, pubchem11869, s-1, 2, 3, 4-tetrahydronaphthoic acid, 1s-1,2,3,4-tetrahydro-1-naphthoic acid
PubChem CID 24820460
IUPAC Name (1S)-1,2,3,4-tetrahydronaphthalene-1-carboxylic acid
SMILES C1CC(C2=CC=CC=C2C1)C(=O)O

Specifications

Melting Point 55°C to 57°C
Quantity 250 mg
Optical Rotation −12° (c=1 in Ethanol)
Formula Weight 176.22
Percent Purity 98%
Chemical Name or Material (S)-1,2,3,4-Tetrahydro-1-naphthoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only