CAS RN 85977-52-2
IUPAC Name:
1,2,3,4-tetrahydronaphthalene-1-carboxylic acid
Synonyms:
1,2,3,4-tetrahydronaphthalene-1-carboxylic acid
Molecular Weight (g/mol):
176.22
Molecular Formula:
C11H12O2
InChi Key:
VDLWTJCSPSUGOA-UHFFFAOYNA-N
SMILES:
OC(=O)C1CCCC2=CC=CC=C12
1
–
1
of
1
results
1
Promotions Available
(S)-1,2,3,4-Tetrahydro-1-naphthoic acid, 98%
(S)-1,2,3,4-Tetrahydro-1-naphthoic acid, 98%
CAS: 85977-52-2 Molecular Formula: C11H12O2 Molecular Weight (g/mol): 176.215 MDL Number: MFCD07368365 InChI Key: VDLWTJCSPSUGOA-JTQLQIEISA-N Synonym: s-1,2,3,4-tetrahydro-1-naphthoic acid,s-1,2,3,4-tetrahedro-naphthoic acid,1s-1,2,3,4-tetrahydronaphthalene-1-carboxylic acid,s-1,2,3,4-tetrahydro-naphthoic acid,1s-tetralin-1-carboxylic acid,s---1,2,3,4-tetrahydronaphthoic acid,s-1,2,3,4-tetrahydronaphthalene-1-carboxylic acid,pubchem11869,s-1, 2, 3, 4-tetrahydronaphthoic acid,1s-1,2,3,4-tetrahydro-1-naphthoic acid PubChem CID: 24820460 IUPAC Name: (1S)-1,2,3,4-tetrahydronaphthalene-1-carboxylic acid SMILES: C1CC(C2=CC=CC=C2C1)C(=O)O