missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Ruxolitinib phosphate
Description
Ruxolitinib phosphate is a phosphate salt of Ruxolitinib (Cat. No. 7064). Potent and selective JAK1/JAK2 inhibitor; orally bioavailable.
Specifications
Specifications
| Chemical Name or Material | (3 R)-3-Cyclopentyl-3-[4-(7 H-pyrrolo[2,3-d]pyrimidin-4-yl)-1 H-pyrazol-1-yl]propanenitrile phosphate |
| CAS | 1092939-17-7 |
| Quantity | 50 mg |
| Target | JAK Kinase Inhibitors |
| Molecular Formula | C17H18N6.H3O4P |
| Formula Weight | 2323 |
| Purity | 98% |