Learn More
Ruthenium, 5% on alumina powder, Thermo Scientific Chemicals
Hydrogenation of Aliphatic Carbonyls and Aromatic Rings, Especially Bulky Molecules.
Supplier: Thermo Scientific Chemicals 01174914
Description
Hydrogenation of aliphatic carbonyls and aromatic rings, especially bulky molecules. Ruthenium on alumina catalyst plays an important role for copper-free sonogashira coupling reactions. It is likewise a useful catalyst for the racemization of secondary alcohols.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Ruthenium | |
| Dark powder | |
| Odorless | |
| Ru | |
| on carbon, rutenio, black, unii-7ui0tkc3u5, 7ui0tkc3u5, ruthenium, powder, alumina, atom, rutherium black, ruthenium/carbon | |
| KJTLSVCANCCWHF-UHFFFAOYSA-N | |
| ruthenium | |
| 23950 |
| 7440-18-8 | |
| 25 g | |
| Powder | |
| MFCD00011207 MFCD03458417 | |
| Insoluble in water. | |
| [Ru] | |
| 101.07 | |
| CHEBI:30682 |
Chemical Identifiers
| 7440-18-8 | |
| 101.07 | |
| KJTLSVCANCCWHF-UHFFFAOYSA-N | |
| 23950 | |
| ruthenium |
| Ru | |
| MFCD00011207 MFCD03458417 | |
| on carbon, rutenio, black, unii-7ui0tkc3u5, 7ui0tkc3u5, ruthenium, powder, alumina, atom, rutherium black, ruthenium/carbon | |
| CHEBI:30682 | |
| [Ru] |
Safety and Handling
H335: May cause respiratory irritation.
EINECSNumber : 231-127-1
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only