missing translation for 'onlineSavingsMsg'
Learn More

Ruthenium, 5% on alumina powder, Thermo Scientific Chemicals

Catalog No. p-4408155
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1174906 5 g
AA1174914 25 g
AA1174922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1174906 Supplier Thermo Scientific Chemicals Supplier No. 01174906
Only null left

Hydrogenation of Aliphatic Carbonyls and Aromatic Rings, Especially Bulky Molecules.

Hydrogenation of aliphatic carbonyls and aromatic rings, especially bulky molecules. Ruthenium on alumina catalyst plays an important role for copper-free sonogashira coupling reactions. It is likewise a useful catalyst for the racemization of secondary alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-18-8
Molecular Formula Ru
Molecular Weight (g/mol) 101.07
MDL Number MFCD00011207 MFCD03458417
InChI Key KJTLSVCANCCWHF-UHFFFAOYSA-N
Synonym on carbon, rutenio, black, unii-7ui0tkc3u5, 7ui0tkc3u5, ruthenium, powder, alumina, atom, rutherium black, ruthenium/carbon
PubChem CID 23950
ChEBI CHEBI:30682
IUPAC Name ruthenium
SMILES [Ru]

Specifications

Appearance Dark powder
Quantity 5 g
Odor Odorless
Form Powder
Solubility Information Insoluble in water.
Chemical Name or Material Ruthenium
Hazard Category H335: May cause respiratory irritation.
EINECSNumber 231-127-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only