missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Rutaecarpine

Chemical Identifiers

CAS 84-26-4
Chemical Name or Material Rutaecarpine
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AC465980010
View Documents Promotion Details
Thermo Scientific™
465980010
1 g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC465982500
View Documents Promotion Details
Thermo Scientific™
465982500
250 mg N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

84-26-4
Rutaecarpine

Specifications

84-26-4
Solubility aq. soln.: pracitically insoluble. Other solubilities: soluble in alcohol, benzene, chloroform, ether
259.0°C to 260.0°C
Rutaecarpine
Videos
SDS
Documents

Documents

RUO – Research Use Only