missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Rufinamide

Catalog No. 382850
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
382850 50 mg
382810 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 382850 Supplier R&D Systems™ Supplier No. 3828/50*

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

A Tocris product, part of R&D Systems. Prolongs inactivation of sodium channels; anticonvulsant

Board spectrum anticonvulsant. Prolongs the inactivation of sodium channels and limits the frequency of action potential firing in cultured and acutely isolated neurons. Displays anticonvulsive activity in a range of animal seizure models.

Chemical Identifiers

CAS 106308-44-5
Molecular Formula C10H8F2N4O

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 238.19
Purity >99%
Molecular Weight (g/mol) 238.198
Chemical Name or Material Rufinamide
Recommended Storage Desiccate at Room Temperature