missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RuCl2[(S)-xylbinap][(S,S)-dpen], TCI America™
Supplier: TCI America R01341G
Specifications
| RuCl2[(S)-xylbinap][(S,S)-dpen] | |
| C66H64Cl2N2P2Ru | |
| 1 g | |
| HYMSONXJNGZZBM-LISIALKWSA-L | |
| [1-[2-bis(3,5-dimethylphenyl)phosphanylnaphthalen-1-yl]naphthalen-2-yl]-bis(3,5-dimethylphenyl)phosphane;dichlororuthenium;(1S,2S)-1,2-diphenylethane-1,2-diamine | |
| 11136945 | |
| Crystalline Powder |
| 220114-03-4 | |
| MFCD09753030 | |
| Dichloro[(S)-2,2′C-bis[di(3,5-xylyl)phosphino]-1,1′C-binaphthyl][(S,S)-1,2-diphenylethylenediamine]ruthenium(II) | |
| CC1=CC(=CC(=C1)P(C2=C(C3=CC=CC=C3C=C2)C4=C(C=CC5=CC=CC=C54)P(C6=CC(=CC(=C6)C)C)C7=CC(=CC(=C7)C)C)C8=CC(=CC(=C8)C)C)C.C1=CC=C(C=C1)C(C(C2=CC=CC=C2)N)N.Cl[Ru]Cl | |
| 1119.17 | |
| 1119.17 |
Chemical Identifiers
| 220114-03-4 | |
| 1119.17 | |
| HYMSONXJNGZZBM-LISIALKWSA-L | |
| 11136945 | |
| CC1=CC(=CC(=C1)P(C2=C(C3=CC=CC=C3C=C2)C4=C(C=CC5=CC=CC=C54)P(C6=CC(=CC(=C6)C)C)C7=CC(=CC(=C7)C)C)C8=CC(=CC(=C8)C)C)C.C1=CC=C(C=C1)C(C(C2=CC=CC=C2)N)N.Cl[Ru]Cl |
| C66H64Cl2N2P2Ru | |
| MFCD09753030 | |
| Dichloro[(S)-2,2′C-bis[di(3,5-xylyl)phosphino]-1,1′C-binaphthyl][(S,S)-1,2-diphenylethylenediamine]ruthenium(II) | |
| [1-[2-bis(3,5-dimethylphenyl)phosphanylnaphthalen-1-yl]naphthalen-2-yl]-bis(3,5-dimethylphenyl)phosphane;dichlororuthenium;(1S,2S)-1,2-diphenylethane-1,2-diamine |
Safety and Handling
TSCA : No