missing translation for 'onlineSavingsMsg'
Learn More
Learn More
[RuCl(p-cymene)((S)-tolbinap)]Cl, TCI America™
Supplier: TCI America R01491G
Specifications
| [RuCl(p-cymene)((S)-tolbinap)]Cl | |
| C58H54Cl2P2Ru | |
| 1 g | |
| BHIUOOHVIWKUTN-UHFFFAOYSA-L | |
| [1-[2-bis(4-methylphenyl)phosphanylnaphthalen-1-yl]naphthalen-2-yl]-bis(4-methylphenyl)phosphane;1-methyl-4-propan-2-ylbenzene;ruthenium(2+);dichloride | |
| 91972163 | |
| Crystalline Powder |
| 228120-95-4 | |
| MFCD09753014 | |
| Chloro[(S)-(-)-2,2′C-bis(di-p-tolylphosphino)-1,1′C-binaphthyl](p-cymene)ruthenium(II) Chloride | |
| CC1=CC=C(C=C1)C(C)C.CC1=CC=C(C=C1)P(C2=CC=C(C=C2)C)C3=C(C4=CC=CC=C4C=C3)C5=C(C=CC6=CC=CC=C65)P(C7=CC=C(C=C7)C)C8=CC=C(C=C8)C.[Cl-].[Cl-].[Ru+2] | |
| 984.988 | |
| 984.99 |
Chemical Identifiers
| 228120-95-4 | |
| 984.988 | |
| BHIUOOHVIWKUTN-UHFFFAOYSA-L | |
| 91972163 | |
| CC1=CC=C(C=C1)C(C)C.CC1=CC=C(C=C1)P(C2=CC=C(C=C2)C)C3=C(C4=CC=CC=C4C=C3)C5=C(C=CC6=CC=CC=C65)P(C7=CC=C(C=C7)C)C8=CC=C(C=C8)C.[Cl-].[Cl-].[Ru+2] |
| C58H54Cl2P2Ru | |
| MFCD09753014 | |
| Chloro[(S)-(-)-2,2′C-bis(di-p-tolylphosphino)-1,1′C-binaphthyl](p-cymene)ruthenium(II) Chloride | |
| [1-[2-bis(4-methylphenyl)phosphanylnaphthalen-1-yl]naphthalen-2-yl]-bis(4-methylphenyl)phosphane;1-methyl-4-propan-2-ylbenzene;ruthenium(2+);dichloride |
Safety and Handling
TSCA : No