missing translation for 'onlineSavingsMsg'
Learn More

[RuCl(p-cymene)((S)-tolbinap)]Cl, TCI America™

Catalog No. p-7135728
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R0149200MG 200 mg
R01491G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R0149200MG Supplier TCI America Supplier No. R0149200MG
Only null left

Chemical Identifiers

CAS 228120-95-4
Molecular Formula C58H54Cl2P2Ru
Molecular Weight (g/mol) 984.988
MDL Number MFCD09753014
InChI Key BHIUOOHVIWKUTN-UHFFFAOYSA-L
Synonym Chloro[(S)-(-)-2,2′C-bis(di-p-tolylphosphino)-1,1′C-binaphthyl](p-cymene)ruthenium(II) Chloride
PubChem CID 91972163
IUPAC Name [1-[2-bis(4-methylphenyl)phosphanylnaphthalen-1-yl]naphthalen-2-yl]-bis(4-methylphenyl)phosphane;1-methyl-4-propan-2-ylbenzene;ruthenium(2+);dichloride
SMILES CC1=CC=C(C=C1)C(C)C.CC1=CC=C(C=C1)P(C2=CC=C(C=C2)C)C3=C(C4=CC=CC=C4C=C3)C5=C(C=CC6=CC=CC=C65)P(C7=CC=C(C=C7)C)C8=CC=C(C=C8)C.[Cl-].[Cl-].[Ru+2]

Specifications

Quantity 200 mg
Formula Weight 984.99
Physical Form Crystalline Powder
Chemical Name or Material [RuCl(p-cymene)((S)-tolbinap)]Cl