missing translation for 'onlineSavingsMsg'
Learn More

[RuCl(p-cymene)((S)-segphos(regR))]Cl, TCI America™

Catalog No. R0155200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R0155200MG 200 mg
R01551G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R0155200MG Supplier TCI America Supplier No. R0155200MG
Only null left

Chemical Identifiers

CAS 944451-29-0
Molecular Formula C48H42Cl2O4P2Ru
Molecular Weight (g/mol) 916.778
MDL Number MFCD09753017
InChI Key HZLSGDCLBWZKRB-UHFFFAOYSA-L
Synonym Chloro[(S)-(-)-5,5′C-bis(diphenylphosphino)-4,4′C-bi-1,3-benzodioxole](p-cymene)ruthenium(II) Chloride
PubChem CID 72376325
IUPAC Name dichlororuthenium;[4-(5-diphenylphosphanyl-1,3-benzodioxol-4-yl)-1,3-benzodioxol-5-yl]-diphenylphosphane;1-methyl-4-propan-2-ylbenzene
SMILES CC1=CC=C(C=C1)C(C)C.C1OC2=C(O1)C(=C(C=C2)P(C3=CC=CC=C3)C4=CC=CC=C4)C5=C(C=CC6=C5OCO6)P(C7=CC=CC=C7)C8=CC=CC=C8.Cl[Ru]Cl

Specifications

Quantity 200 mg
Formula Weight 916.78
Physical Form Crystalline Powder
Chemical Name or Material [RuCl(p-cymene)((S)-segphos(regR))]Cl