accessibility menu, dialog, popup

UserName

CAS RN 944451-29-0

CH3CH3H3COOPOOPClRuCl(?)

CAS RN 944451-29-0

IUPAC Name: 1-methyl-4-(propan-2-yl)benzene; dichlororuthenium; {4-[5-(diphenylphosphanyl)-2H-1,3-benzodioxol-4-yl]-2H-1,3-benzodioxol-5-yl}diphenylphosphane
Synonyms: cymene; dichlororuthenium; segphos
Molecular Weight (g/mol): 916.78
Molecular Formula: C48H42Cl2O4P2Ru
InChi Key: HZLSGDCLBWZKRB-UHFFFAOYSA-L
SMILES: Cl[Ru]Cl.CC(C)C1=CC=C(C)C=C1.C1OC2=C(O1)C(=C(C=C2)P(C1=CC=CC=C1)C1=CC=CC=C1)C1=C(C=CC2=C1OCO2)P(C1=CC=CC=C1)C1=CC=CC=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results