missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ru-MACHO(regR) (contains 5% Toluene at maximum), TCI America™
Supplier: TCI America R01361G
Specifications
| Ru-MACHO(regR) (contains 5% Toluene at maximum) | |
| White-Yellow | |
| MFCD22377801 | |
| Carbonylchlorohydrido[bis(2-diphenylphosphinoethyl)amino]ruthenium(II) | |
| [C-]#[O+].C1=CC=C(C=C1)P(CCNCCP(C2=CC=CC=C2)C3=CC=CC=C3)C4=CC=CC=C4.Cl[RuH] | |
| 607.033 | |
| 607.03 |
| 1295649-40-9 | |
| C29H30ClNOP2Ru | |
| 1 g | |
| YCZWZQUMPDDGQF-UHFFFAOYSA-M | |
| carbon monoxide;chloro(hydrido)ruthenium;2-diphenylphosphanyl-N-(2-diphenylphosphanylethyl)ethanamine | |
| 132774591 | |
| Crystalline Powder |
Chemical Identifiers
| 1295649-40-9 | |
| 607.033 | |
| YCZWZQUMPDDGQF-UHFFFAOYSA-M | |
| 132774591 | |
| [C-]#[O+].C1=CC=C(C=C1)P(CCNCCP(C2=CC=CC=C2)C3=CC=CC=C3)C4=CC=CC=C4.Cl[RuH] |
| C29H30ClNOP2Ru | |
| MFCD22377801 | |
| Carbonylchlorohydrido[bis(2-diphenylphosphinoethyl)amino]ruthenium(II) | |
| carbon monoxide;chloro(hydrido)ruthenium;2-diphenylphosphanyl-N-(2-diphenylphosphanylethyl)ethanamine |
Safety and Handling
TSCA : No