missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ru-MACHO(regR) (contains 5% Toluene at maximum), TCI America™
$121.16 - $378.40
Chemical Identifiers
| CAS | 1295649-40-9 |
|---|---|
| Molecular Formula | C29H30ClNOP2Ru |
| Molecular Weight (g/mol) | 607.033 |
| MDL Number | MFCD22377801 |
| InChI Key | YCZWZQUMPDDGQF-UHFFFAOYSA-M |
| Synonym | Carbonylchlorohydrido[bis(2-diphenylphosphinoethyl)amino]ruthenium(II) |
| PubChem CID | 132774591 |
| IUPAC Name | carbon monoxide;chloro(hydrido)ruthenium;2-diphenylphosphanyl-N-(2-diphenylphosphanylethyl)ethanamine |
| SMILES | [C-]#[O+].C1=CC=C(C=C1)P(CCNCCP(C2=CC=CC=C2)C3=CC=CC=C3)C4=CC=CC=C4.Cl[RuH] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
R0136200MG
|
TCI America
R0136200MG |
200 mg |
Each for $121.16
|
|
|||||
|
R01361G
|
TCI America
R01361G |
1 g |
Each for $378.40
|
|
|||||
Chemical Identifiers
| 1295649-40-9 | |
| 607.033 | |
| YCZWZQUMPDDGQF-UHFFFAOYSA-M | |
| 132774591 | |
| [C-]#[O+].C1=CC=C(C=C1)P(CCNCCP(C2=CC=CC=C2)C3=CC=CC=C3)C4=CC=CC=C4.Cl[RuH] |
| C29H30ClNOP2Ru | |
| MFCD22377801 | |
| Carbonylchlorohydrido[bis(2-diphenylphosphinoethyl)amino]ruthenium(II) | |
| carbon monoxide;chloro(hydrido)ruthenium;2-diphenylphosphanyl-N-(2-diphenylphosphanylethyl)ethanamine |
Specifications
| 1295649-40-9 | |
| C29H30ClNOP2Ru | |
| 200 mg | |
| YCZWZQUMPDDGQF-UHFFFAOYSA-M | |
| carbon monoxide;chloro(hydrido)ruthenium;2-diphenylphosphanyl-N-(2-diphenylphosphanylethyl)ethanamine | |
| 132774591 | |
| Crystalline Powder |
| White-Yellow | |
| MFCD22377801 | |
| Carbonylchlorohydrido[bis(2-diphenylphosphinoethyl)amino]ruthenium(II) | |
| [C-]#[O+].C1=CC=C(C=C1)P(CCNCCP(C2=CC=CC=C2)C3=CC=CC=C3)C4=CC=CC=C4.Cl[RuH] | |
| 607.033 | |
| 607.03 | |
| Ru-MACHO(regR) (contains 5% Toluene at maximum) |
Safety and Handling
TSCA : No