accessibility menu, dialog, popup

UserName

COPNHPClRu

CAS RN 1295649-40-9

IUPAC Name: bis[2-(diphenylphosphanyl)ethyl]amine; chloro(hydrido)ruthenium; methanidylidyneoxidanium
Synonyms: bis[2-(diphenylphosphanyl)ethyl]amine; carbon monoxide; chloro(hydrido)ruthenium
Molecular Weight (g/mol): 607.03
Molecular Formula: C29H30ClNOP2Ru
InChi Key: YCZWZQUMPDDGQF-UHFFFAOYSA-M
SMILES: Cl[RuH].[C-]#[O+].C(CP(C1=CC=CC=C1)C1=CC=CC=C1)NCCP(C1=CC=CC=C1)C1=CC=CC=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results