missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Rotigotine

Catalog No. AC466541000
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC466541000 100 mg
AC466545000 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC466541000 Supplier Thermo Scientific Chemicals Supplier No. 466541000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 99755-59-6 | C19H25NOS, C19H25NOS | 315.48 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 99755-59-6
Molecular Formula C19H25NOS

Specifications

Quantity 100 mg
Formula Weight 315.47
InChI Key KFQYTPMOWPVWEJ-UHFFFAOYNA-N
SMILES CCCN(CCC1=CC=CS1)C1CCC2=C(O)C=CC=C2C1
IUPAC Name 6-{propyl[2-(thiophen-2-yl)ethyl]amino}-5,6,7,8-tetrahydronaphthalen-1-ol
Molecular Weight (g/mol) 315.48
Chemical Name or Material Rotigotine
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Harmful if swallowed.

Precautionary Statement GHS P Statement IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Call a POISON CENTER or doctor/physician if you feel unwell.

Wash face, hands and any exposed skin thoroughly after handling.

RUO – Research Use Only