Learn More
Thermo Scientific Chemicals Rosolic acid
CAS: 603-45-2 | C19H14O3 | 290.32 g/mol
Supplier: Thermo Scientific Chemicals 229830250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
IndicatorSpecifications
| Rosolic acid | |
| Brown-Red | |
| Authentic | |
| C19H14O3 | |
| 25 g | |
| 15,872 | |
| Solubility in water: practically insoluble. Other solubilities: freely soluble in naoh,koh,70% h2so4 and hcl,freely soluble in alcohol,slightly soluble in ether and chloroform | |
| OC1=CC=C(C=C1)C(C1=CC=C(O)C=C1)=C1C=CC(=O)C=C1 | |
| 290.32 | |
| CHEBI:34544 | |
| Crystalline Powder |
| 603-45-2 | |
| 10% max. (110°C, 1 hr) | |
| Glass bottle | |
| MFCD00001624 | |
| 08,361 | |
| aurin, rosolic acid, corallin, p-rosolic acid, aurine, spirit aurine, corallin spirit soluble, 4,4'-dihydroxyfuchsone, aurin no. 555, 4-bis 4-hydroxyphenyl methylene cyclohexa-2,5-dienone | |
| FYEHYMARPSSOBO-UHFFFAOYSA-N | |
| 4-[bis(4-hydroxyphenyl)methylidene]cyclohexa-2,5-dien-1-one | |
| 5100 | |
| 290.32 |
Chemical Identifiers
| 603-45-2 | |
| 290.32 | |
| FYEHYMARPSSOBO-UHFFFAOYSA-N | |
| 5100 | |
| 4-[bis(4-hydroxyphenyl)methylidene]cyclohexa-2,5-dien-1-one |
| C19H14O3 | |
| MFCD00001624 | |
| aurin, rosolic acid, corallin, p-rosolic acid, aurine, spirit aurine, corallin spirit soluble, 4,4'-dihydroxyfuchsone, aurin no. 555, 4-bis 4-hydroxyphenyl methylene cyclohexa-2,5-dienone | |
| CHEBI:34544 | |
| OC1=CC=C(C=C1)C(C1=CC=C(O)C=C1)=C1C=CC(=O)C=C1 |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee
GHS Signal Word: Warning
EINECSNumber : 210-041-8
TSCA : TSCA
RUO – Research Use Only