missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Rosolic acid

Catalog No. p-191138
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC229830250 25 g Glass bottle
AC229831000 100 g Glass bottle
AC229835000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC229830250 Supplier Thermo Scientific Chemicals Supplier No. 229830250
Only null left

CAS: 603-45-2 | C19H14O3 | 290.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator

Chemical Identifiers

CAS 603-45-2
Molecular Formula C19H14O3
Molecular Weight (g/mol) 290.32
MDL Number MFCD00001624
InChI Key FYEHYMARPSSOBO-UHFFFAOYSA-N
Synonym aurin, rosolic acid, corallin, p-rosolic acid, aurine, spirit aurine, corallin spirit soluble, 4,4'-dihydroxyfuchsone, aurin no. 555, 4-bis 4-hydroxyphenyl methylene cyclohexa-2,5-dienone
PubChem CID 5100
ChEBI CHEBI:34544
IUPAC Name 4-[bis(4-hydroxyphenyl)methylidene]cyclohexa-2,5-dien-1-one
SMILES OC1=CC=C(C=C1)C(C1=CC=C(O)C=C1)=C1C=CC(=O)C=C1

Specifications

Color Brown-Red
Loss on Drying 10% max. (110°C, 1 hr)
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 25 g
Beilstein 08,361
Merck Index 15,872
Solubility Information Solubility in water: practically insoluble. Other solubilities: freely soluble in naoh,koh,70% h2so4 and hcl,freely soluble in alcohol,slightly soluble in ether and chloroform
Formula Weight 290.32
Physical Form Crystalline Powder
Chemical Name or Material Rosolic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee
EINECSNumber 210-041-8
TSCA TSCA

RUO – Research Use Only